Order Entry
Austria
Orders LinkContactUsLinkComponent
Charts

Charts

C479, +5...+40 °C; 0 - 100 % RH , 7 days
Catalog # 620-0418
Supplier:   DICKSON
Charts
Catalog # 620-0418
Supplier:  DICKSON
Charts

C479, +5...+40 °C; 0 - 100 % RH , 7 days

Price
79,10 €
Log in for availability
1 * 60 ST
Product Details & Documents

Documents

Specifications
  • Description:
    C479
  • Seq:
    50
  • Range:
    +5...+40 °C; 0 - 100 % RH
  • Recording time:
    7 days
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.