Order Entry
Austria
Orders LinkContactUsLinkComponent
Charts

Charts

C476, +40...+110 °C; 0 - 100 % RH, 24 hours
Catalog # 620-0415
Supplier:   DICKSON
Charts
Catalog # 620-0415
Supplier:  DICKSON
Charts

C476, +40...+110 °C; 0 - 100 % RH, 24 hours

Price
79,10 €
Log in for availability
1 * 60 ST
Product Details & Documents

Documents

Specifications
  • Description:
    C476
  • Seq:
    60
  • Range:
    +40...+110 °C; 0 - 100 % RH
  • Recording time:
    24 hours
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.