Charts
C415, –20...+120 °F; 0 - 100 % RH, 24 hours
Artikelnr. 620-0411
Anbieter: DICKSON
Jump to:
Product Details & Documents
Specifications
- Description:C415
- Seq:17
- Range:–20...+120 °F; 0 - 100 % RH
- Recording time:24 hours
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



