Orders LinkContactUsLinkComponent
Accessories for screw thread joints
Accessories for screw thread joints
6,0 -7,0 mm, 24
Catalog # BIBBQW24/6
Supplier:   SciLabware
Accessories for screw thread joints
Catalog # BIBBQW24/6
Supplier:  SciLabware
Accessories for screw thread joints
Accessories for screw thread joints
6,0 -7,0 mm, 24
Price
40,30 €
Log in for availability
1 * 10 ST
Product Details & Documents
For use with Quickfit® glass screw thread.
Specifications
  • Hole Ø:
    6,0 -7,0 mm
  • Thread:
    24