Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Unisette™, M505 | Avantor

Tissue processing/embedding cassettes, Unisette™, M505

Turquoise
Artikelnr. 720-1552
Anbieter:   Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505
Artikelnr. 720-1552
Anbieter:  Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505

Turquoise

Preis
231,00 €
Einloggen für Verfügbarkeit
1 * 1.500 ST
Product Details & Documents

Made from high density polymer. These cassettes, with lids, keep specimens safe whilst submerged in liquid.

  • Flow-through slots maximise fluid exchange and ensure effective drainage
  • Resistant to histological solvents
  • Integral lid
  • Front writing face inclined 35°
  • Available in 11 non cytotoxic, non metallic colours, as well as three fluorescent colours

Verpackung: Each case contains three boxes of 500 cassettes.

Spezifikationen
  • Colour:
    Turquoise
  • Seq:
    13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.