Tissue processing/embedding cassettes, Unisette™, M505
Yellow
Catalog # 720-1559
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505
Catalog # 720-1559
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Made from high density polymer. These cassettes, with lids, keep specimens safe whilst submerged in liquid.
- Flow-through slots maximise fluid exchange and ensure effective drainage
- Resistant to histological solvents
- Integral lid
- Front writing face inclined 35°
- Available in 11 non cytotoxic, non metallic colours, as well as three fluorescent colours
Verpackung: Each case contains three boxes of 500 cassettes.
Specifications
- Colour:Yellow
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



