Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Unisette™, M505

Tissue processing/embedding cassettes, Unisette™, M505

Green
Catalog # 720-1558
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505
Catalog # 720-1558
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505

Green

Price
231,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

Made from high density polymer. These cassettes, with lids, keep specimens safe whilst submerged in liquid.

  • Flow-through slots maximise fluid exchange and ensure effective drainage
  • Resistant to histological solvents
  • Integral lid
  • Front writing face inclined 35°
  • Available in 11 non cytotoxic, non metallic colours, as well as three fluorescent colours

Packaging: Each case contains three boxes of 500 cassettes.

Specifications
  • Colour:
    Green
  • Seq:
    03