Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Unisette™, M505

Tissue processing/embedding cassettes, Unisette™, M505

Pink
Catalog # 720-1557
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505
Catalog # 720-1557
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Unisette™, M505

Pink

Price
231,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

Made from high density polymer. These cassettes, with lids, keep specimens safe whilst submerged in liquid.

  • Flow-through slots maximise fluid exchange and ensure effective drainage
  • Resistant to histological solvents
  • Integral lid
  • Front writing face inclined 35°
  • Available in 11 non cytotoxic, non metallic colours, as well as three fluorescent colours

Packaging: Each case contains three boxes of 500 cassettes.

Specifications
  • Colour:
    Pink
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.