Orders LinkContactUsLinkComponent
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
Biowrap 60x60 mm, white
Catalog # 720-2355
Supplier:   CELLPATH
Biopsy specimen processing, CellSafe™
Catalog # 720-2355
Supplier:  CELLPATH
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
Biowrap 60x60 mm, white
Price
25,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
  • No tissue damage
  • Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
Specifications
  • Description:
    Biowrap 60x60 mm, white