Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Application specific filter papers, grade 72, Whatman™

72, 50 mm
  WHAT1872-050
Application specific filter papers, grade 72, Whatman™
  WHAT1872-050
Application specific filter papers, grade 72, Whatman™
Application specific filter papers, grade 72, Whatman™

72, 50 mm

 
202,00 €
   
1 * 100 ST
 

Grade 72 activated carbon air filter paper is designed for air pollution monitoring of radioactive iodine in medical and nuclear installations.

  • High-quality composite cellulose/glass filter paper
  • Loaded with activated carbon for radioactive iodine absorption
  • Suitable for use in radioactive environments

Air pollution monitoring of radioactive iodine in medical or nuclear installations is a critical safety requirement. Grade 72 cellulose filter papers are designed specifically for these environments, delivering sensitive and accurate results by absorbing any radioactive iodine in the air for analysis.

 
  • Grade:
    72
  • Seq:
    02
  • Size:
    50 mm
  • Thickness:
    800 µm
  • Cat. No.:
    WHAT1872-050
  • Weight:
    195 g/m²