Orders LinkContactUsLinkComponent
Inoculating loops, disposable, Microloops
Inoculating loops, disposable, Microloops
Loop peel pouch, 1 µl, Blue
Catalog # 300-0511
Inoculating loops, disposable, Microloops
Catalog # 300-0511
Inoculating loops, disposable, Microloops
Inoculating loops, disposable, Microloops
Loop peel pouch, 1 µl, Blue
Price
339,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Sterile, disposable inoculating loops.
  • Colour coded plastic
  • Moderately flexible
Specifications
  • Capacity:
    1 µl
  • Colour:
    Blue
  • Description:
    Loop peel pouch