Order Entry
Austria
Orders LinkContactUsLinkComponent
Flat optical windows

Flat optical windows

Flat optics disc, Quartz (Suprasil®), 22 mm, 1,25 mm
Catalog # HELL202-40
Supplier:   Hellma
Flat optical windows
Catalog # HELL202-40
Supplier:  Hellma
Flat optical windows
Flat optical windows

Flat optics disc, Quartz (Suprasil®), 22 mm, 1,25 mm

Price
38,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Optical glass cells and elements dedicated to special applications with detachable window.

Suitable for high precision analytical techniques.

Specifications
  • Window material:
    Quartz (Suprasil®)
  • Description:
    Flat optics disc
  • Dimensions:
    22 mm
  • Thickness:
    1,25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.