Filtration funnels
20 ml, 130 mm, 75 mm, 4, 8 mm, 65 mm
Catalog # BIBB3730/08
Supplier: SciLabware
Jump to:
Product Details & Documents
Pyrex® borosilicate glass.
- Sintered glass disc
- Filter paper is not required for filtration of solutions
- Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
Specifications
- Porosity:4
- Capacity:20 ml
- Length:130 mm
- Int.Ø top:65 mm
- Stem O-Ø:8 mm
- Stem length:75 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



