Order Entry
Austria
Orders LinkContactUsLinkComponent
Desiccators, PYREX®

Desiccators, PYREX®

Desiccator, Non-vacuum, for use at atmospheric pressure, with knob, 150 DN, 85 mm, 215×154 mm, 170 mm
  BIBB1590/02D
 :   SciLabware
Desiccators, PYREX®
  BIBB1590/02D
Desiccators, PYREX®
Desiccators, PYREX®

Desiccator, Non-vacuum, for use at atmospheric pressure, with knob, 150 DN, 85 mm, 215×154 mm, 170 mm

 
200,00 €
   
1 * 1 ST
 

Ideal for vacuum and non vacuum applications.

  • Ground glass flanges
  • Borosilicate glass base and lid
  • Plates positioned on an internal ledge within the base
 
  • Ø int.:
    170 mm
  • Nominal size:
    150 DN
  • Type:
    Non-vacuum, for use at atmospheric pressure, with knob
  • Description:
    Desiccator
  • Seq:
    001
  • Usable height:
    85 mm
  • Ø ext.×H:
    215×154 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.