Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Pasteur pipettes

Pasteur pipette, graduated, 1 ml, 153 mm, -, 500 pieces/carton
  GREI612302_500
Pasteur pipettes
  GREI612302_500
Pasteur pipettes

Pasteur pipette, graduated, 1 ml, 153 mm, -, 500 pieces/carton

 
63,20 €
   
1 * 500 ST
 

These PE pipettes are suitable for the rapid transfer of liquids.

 
  • Packed:
    500 pieces/carton
  • Sterile:
    -
  • Capacity:
    1 ml
  • Description:
    Pasteur pipette, graduated
  • Length:
    153 mm
  • Seq:
    02