0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
Orders Link
ContactUsLinkComponent
Buffers
pH Calibration Buffers
Buffer solutions
Buffer solutions
Buffer solution, 12,00, ±0,05, 20 °C, Plastic bottle, 500 mL
REGE11205
:
REAGECON
Buffer solutions
REGE11205
:
REAGECON
:
Buffer solutions
Buffer solution, 12,00, ±0,05, 20 °C, Plastic bottle, 500 mL
:
31,90 €
1 * 500 mL
-
+
Pack type:
Plastic bottle
Pk:
500 mL
Reference temperature:
20 °C
Description:
Buffer solution
pH accuracy:
±0,05
pH value:
12,00
VWR®, General purpose viscosity standards
#: 99445.260
: VWR Chemicals