Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Formaldehyde 7,5%, TECHNICAL, buffered (pH 7.0 ±0.2)

Plastic drum, 10 L
Catalog # 5534.9010
Health
Irritant
Supplier:   VWR Chemicals
Formaldehyde 7,5%, TECHNICAL, buffered (pH 7.0 ±0.2)
Catalog # 5534.9010
Supplier:  VWR Chemicals
CAS Number:  50-00-0
Formaldehyde 7,5%, TECHNICAL, buffered (pH 7.0 ±0.2)
Formaldehyde 7,5%, TECHNICAL, buffered (pH 7.0 ±0.2)

Plastic drum, 10 L

CAS Number:  50-00-0
Price
188,00 €
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specification Test Results

  • Assay (Formaldehyde):
    7 - 9 %
  • pH (20°C):
    6.8 - 7.2

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.