Handheld refractometers, Eclipse
Handheld refractometer for low volume (< 1 µl), 45 - 80 °Bx, 0,2 °Bx
Catalog # 635-0293
Jump to:
Discontinued
Product Details & Documents
Handheld refractometers use high precision clear scale, and simple zero adjustment with lock.
- Shaded field of view for distinction
- Anti-roll support and ergonomic design
- Comfortable eyepiece with smooth adjustment
- Easy to clean prism and push-on flap protection
- Unique sample ‘dribble’ feature
Delivery information: Supplied with a soft carrying case and instructions.
Documents
Specifications
- Range:45 - 80 °Bx
- Resolution:0,2 °Bx
- Description:Handheld refractometer for low volume (< 1 µl)
- Seq:16