Order Entry
Austria
Orders LinkContactUsLinkComponent
Handheld refractometers, Eclipse

Handheld refractometers, Eclipse

Handheld refractometer for sugar, 0 - 30 % Bx, 0,2 % Bx
Catalog # 635-0038
Supplier:   BELLINGHAM STANLEY
Handheld refractometers, Eclipse
Catalog # 635-0038
Supplier:  BELLINGHAM STANLEY
Handheld refractometers, Eclipse
Handheld refractometers, Eclipse

Handheld refractometer for sugar, 0 - 30 % Bx, 0,2 % Bx

Price
350,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Handheld refractometers use high precision clear scale, and simple zero adjustment with lock.

  • Shaded field of view for distinction
  • Anti-roll support and ergonomic design
  • Comfortable eyepiece with smooth adjustment
  • Easy to clean prism and push-on flap protection
  • Unique sample ‘dribble’ feature

Delivery information: Supplied with a soft carrying case and instructions.

Specifications
  • Range:
    0 - 30 % Bx
  • Resolution:
    0,2 % Bx
  • Description:
    Handheld refractometer for sugar
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.