Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TAE buffer solution 25X concentrate (TRIS-acetate-EDTA buffer), 1 KIT
Catalog # AMBI9870
Marketsource
Supplier:   Ambion
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # AMBI9870
Supplier:  Ambion
Marketsource
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TAE buffer solution 25X concentrate (TRIS-acetate-EDTA buffer), 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    TAE buffer solution 25X concentrate (TRIS-acetate-EDTA buffer)

Specifications