Orders LinkContactUsLinkComponent
Dissecting needles
Dissecting needles
Straight, 140 mm
Catalog # 233-0102
Supplier:   BOCHEM
Dissecting needles
Catalog # 233-0102
Supplier:  BOCHEM
Dissecting needles
Dissecting needles
Straight, 140 mm
Price
2,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dissecting needles made of stainless steel, with plastic handle.
Specifications
  • Form:
    Straight
  • Length:
    140 mm