Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Amitraz D12 (2,2´,4,4´-tetramethyl D3) 100 ng/µl 1 * 1,1 mL
Catalog # EHERXA10230100AC
Supplier:
EHRENSTORFER
Expand image
Amitraz D12 (2,2´,4,4´-tetramethyl D3) 100 ng/µl 1 * 1,1 mL
Catalog # EHERXA10230100AC
Supplier:
EHRENSTORFER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Standard solutions for analytical quality assurance, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW