Acetochlor D11 100 ng/µl 1 * 1,1 mL
Catalog # EHERXA10018100AC
Supplier: EHRENSTORFER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Butane-1,3-diol, Reference Standard
Supplier: USP
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
Supelco® Organic Reference Standard, beta-Pinene solution
Supplier: Merck
VWR® PESTINORM®, Gas Chromatography (GC) Certified Reference Material
Supplier: VWR Chemicals
Acesulfame Potassium, Reference Standard
Supplier: USP
VWR®, Colour standards, Platinum-Cobalt (Pt-Co, Hazen)
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Sigma-Aldrich colour chart, analytical standard, Supelco®
Supplier: Merck
Buffer PH 4,00 1 * 500 mL
Supplier: REAGECON
Lumefantrine, Reference Standards
Supplier: USP
NAPROXEN IMPURITY N 1 * 100 mg
Supplier: Merck
SECUMATIC FDR 20L 1 * 20 L
Supplier: KAMMERER
Supelco® Organic Reference Standard, Arbutin
Supplier: Merck