Order Entry
Austria
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

J.T.Baker® BAKER ANALYZED®, Buffer Solutions

11,00, ±0,05, Glycine, 20 °C, Plastic bottle, 1 L
Catalog # 7267.1000
Supplier:   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Catalog # 7267.1000
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

11,00, ±0,05, Glycine, 20 °C, Plastic bottle, 1 L

CAS Number:  
Price
70,29 €
Log in for availability
1 * 1 L
Product Details & Documents

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.

Contain 0,01% Sodium benzoate.

 : This buffer is traceable to NIST Standard reference buffer.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Glycine
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0,05
  • pH value:
    11,00

Specifications