Single - component oxygenates solutions
tert-Butanol, with 50000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 mL
Catalog # REST30470
Supplier: Restek
Jump to:
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Pk:1 mL
- Standards:US EPA 502.2, 8021, 8206B
- Description:tert-Butanol, with 50000 µg/ml in purge and trap grade methanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST30470
- Formula:(CH₃)₃COH
- Description:tert-Butanol (TBA)
- CAS No.:75-65-0



