Order Entry
Austria
Orders LinkContactUsLinkComponent
Single - component oxygenates solutions

Single - component oxygenates solutions

Methyl tert-butyl ether (MTBE), with 2000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 mL
Catalog # REST30402
Supplier:   Restek
Single - component oxygenates solutions
Catalog # REST30402
Supplier:  Restek
CAS Number:  
Single - component oxygenates solutions
Single - component oxygenates solutions

Methyl tert-butyl ether (MTBE), with 2000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 mL

CAS Number:  
Price
52,50 €
Log in for availability
1 * 1 mL
Product Details & Documents

Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Pk:
    1 mL
  • Standards:
    US EPA 502.2, 8021, 8206B
  • Description:
    Methyl tert-butyl ether (MTBE), with 2000 µg/ml in purge and trap grade methanol
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST30402
  • Formula:
    (CH₃)₃COCH₃
  • Description:
    Methyl tert-butyl ether (MTBE)
  • CAS No.:
    1634-04-4

Specifications