Orders LinkContactUsLinkComponent
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
24×50 mm, #1,5
Catalog # 631-0147
Supplier:   VWR Collection
VWR® Rectangular Cover Glasses
Catalog # 631-0147
Supplier:  VWR Collection
VWR® Rectangular Cover Glasses
VWR® Rectangular Cover Glasses
24×50 mm, #1,5
Price
89,60 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: In plastic hinged lid boxes.
Specifications
  • No.:
    #1,5
  • L×W:
    24×50 mm
  • Seq:
    13