Orders LinkContactUsLinkComponent
Camera adapters
Camera adapters
C-mount adapter 0,5× for ¹/₃" chip sensors
Catalog # 630-0092
Supplier:   MOTIC
Camera adapters
Catalog # 630-0092
Supplier:  MOTIC
Camera adapters
Camera adapters
C-mount adapter 0,5× for ¹/₃" chip sensors
Price
72,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    C-mount adapter 0,5× for ¹/₃" chip sensors
  • Seq:
    02