Orders LinkContactUsLinkComponent
pH indicator papers, MQuant®, Supelco®
pH indicator papers, MQuant®, Supelco®
Phenolphthalein paper (pH<8,5 colourless / >8,5 red), -, -
Catalog # 1.09521.0003
Supplier:   Merck
pH indicator papers, MQuant®, Supelco®
Catalog # 1.09521.0003
Supplier:  Merck
pH indicator papers, MQuant®, Supelco®
pH indicator papers, MQuant®, Supelco®
Phenolphthalein paper (pH<8,5 colourless / >8,5 red), -, -
Price
61,00 €
Log in for availability
1 * 3 Rolle
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
  • Roll format are easy to use and most common ranges are available in a refill format
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
Specifications
  • pH gradation:
    -
  • pH range:
    -
  • Description:
    Phenolphthalein paper (pH<8,5 colourless / >8,5 red)
  • Cat. No.:
    1.09521.0003
  • M-Clarity quality level:
    MQ100