Orders LinkContactUsLinkComponent
Automatic viscometers, ViscoSystem® AVS® 470
Automatic viscometers, ViscoSystem® AVS® 470
Automatic viscometer with ViscoPump II module for optoelectronic sensing, AVS 470, 0,35 − 1800 (pressure) ; 0,35 − 5000 (suction) mm²/s (cSt)
Catalog # 616-0026
Supplier:   SI Analytics
Automatic viscometers, ViscoSystem® AVS® 470
Catalog # 616-0026
Supplier:  SI Analytics
Automatic viscometers, ViscoSystem® AVS® 470
Automatic viscometers, ViscoSystem® AVS® 470
Automatic viscometer with ViscoPump II module for optoelectronic sensing, AVS 470, 0,35 − 1800 (pressure) ; 0,35 − 5000 (suction) mm²/s (cSt)
Price
11 060,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The ViscoSystem AVS 470 is the first measuring device that allows suction and pressure measurements completely independent of a PC.
  • Determination with pressure or with vacuum for high viscosities
  • Both detections methods optical or thermal are possible
  • Nearly all necessary calculations are integrated
  • Additional parameters can be set up over a regular PS 2 keyboard
This brings maximum independence and flexibility, allowing you to set up a measuring station, that meets highest requirements, even under difficult conditions, to monitor production or control quality in the polymer and mineral oil industry.
The ViscoSystem AVS 470 determines the time which the sample to be examined requires to flow through the measuring distance in the capillary viscometer with quartz precision.
All common types of viscosity calculations are already integrated into the device, a small PS 2 keyboard is all you need to enter additional data.
A serial printer can be used to conveniently document your measuring results.
Specifications
  • Model:
    AVS 470
  • Range:
    0,35 − 1800 (pressure) ; 0,35 − 5000 (suction) mm²/s (cSt)
  • Description:
    Automatic viscometer with ViscoPump II module for optoelectronic sensing
  • Interfaces:
    RS232C for serial printer
  • Cat. No.:
    616-0026
  • Measurement range (flow time):
    <9999,99 s
  • Temperature regulation period:
    0 – 20 minutes
  • W×D×H:
    225×320×205 mm
  • Viscosity range:
    Pressure: 0,35 - 1800 mm²/s (cSt)
    Suction capacity: 0,35 - ~5000 mm²/s (cSt)
  • Pump pressure:
    160 mbar
  • Time measurement accuracy:
    ±0,01 %
  • Weight:
    5,4 kg
  • Display:
    LCD
  • Preselection of the number of measurements:
    Up to 99
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, NBR (Perbunan), VZ 7052
    O-ring seal, NBR (Perbunan), VZ 7052
    SCOI285420577
    Log in for availability
    1 * 1 ST - 21,70 €
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, FKM (viton), VZ 7050
    O-ring seal, FKM (viton), VZ 7050
    SCOI285420552
    Log in for availability
    1 * 1 ST - 22,90 €
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, EPDM (buna), VZ 7051
    O-ring seal, EPDM (buna), VZ 7051
    SCOI285420569
    Log in for availability
    1 * 1 ST - 22,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Safety bottle with glass thread, VZ 7021
    Safety bottle with glass thread, VZ 7021
    SCOI285420269
    Log in for availability
    1 * 1 ST - 133,00 €
Product Information
  • Catalog #
    Availability
    Price
  • VZ 3079 Schlauchkappe, silicon, red, int.Ø 9 mm
    VZ 3079 Schlauchkappe, silicon, red, int.Ø 9 mm
    SCOI285414048
    Log in for availability
    1 * 1 ST - 55,00 €
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470
    VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470
    SCOI285414031
    Log in for availability
    1 * 1 ST - 343,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Ink ribbon VZ 6249
    Ink ribbon VZ 6249
    SCOI285411572
    Log in for availability
    1 * 1 ST - 43,40 €
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5102 Connection cap 'Vacuum', silicon, red, int.Ø 4,5 mm
    VZ 5102 Connection cap 'Vacuum', silicon, red, int.Ø 4,5 mm
    SCOI285410527
    Log in for availability
    1 * 1 ST - 40,90 €
Product Information
  • Catalog #
    Availability
    Price
  • VZ 5101 Connection cap 'Venting', silicon, black, int.Ø 4,5 mm
    VZ 5101 Connection cap 'Venting', silicon, black, int.Ø 4,5 mm
    SCOI285410519
    Log in for availability
    1 * 1 ST - 40,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Thermal paper for Z 900 and Z 910 Z 905
    Thermal paper for Z 900 and Z 910 Z 905
    SCOI285225630
    Log in for availability
    1 * 5 ST - 55,00 €
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, VZ 7053, 8,3×2,4 mm
    O-ring seal, VZ 7053, 8,3×2,4 mm
    SCOI285420585
    Log in for availability
    1 * 1 ST - 189,00 €
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, VZ 7054, 7,0×3,0 mm
    O-ring seal, VZ 7054, 7,0×3,0 mm
    SCOI285420593
    Log in for availability
    1 * 1 ST - 161,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Absorbing filter with activated coal, VZ 7216
    Absorbing filter with activated coal, VZ 7216
    SCOI285422215
    Log in for availability
    1 * 1 ST - 180,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Connectors, VZ 7211, for absorbing filter VZ 7215, 7216
    Connectors, VZ 7211, for absorbing filter VZ 7215, 7216
    SCOI285422167
    Log in for availability
    1 * 1 ST - 94,00 €
Product Information
  • Catalog #
    Availability
    Price
  • O-ring seal, VZ 7055, 5,3×1,8 mm
    O-ring seal, VZ 7055, 5,3×1,8 mm
    SCOI285420606
    Log in for availability
    1 * 1 ST - 117,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Absorbing filter, VZ 7215
    Absorbing filter, VZ 7215
    SCOI285422204
    Log in for availability
    1 * 1 ST - 181,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Dust protection cover, for thermostatic baths
    Dust protection cover, for thermostatic baths
    SCOI285418573
    Log in for availability
    1 * 1 ST - 133,00 €