Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Baumé hydrometers

30 - 70 °Bé, 1, 250 mm
  AMARH843154
 :   Amarell
Baumé hydrometers
  AMARH843154
 :  Amarell
Baumé hydrometers
Baumé hydrometers

30 - 70 °Bé, 1, 250 mm

 
23,20 €
   
1 * 1 ST
 

General purpose Baumé hydrometers with or without thermometer.

  • Reference temperature of 15 °C
  • Paper scale with visible print
  • Shot weighted body

Colourless round capillary with red or blue filling.

 
  • Length:
    250 mm
  • Seq:
    20
  • Division:
    1
  • Range:
    30 - 70 °Bé
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.