Order Entry
Austria
Orders LinkContactUsLinkComponent
Halogen bulbs

Halogen bulbs

Quartz halogen lamp, 6 V, 30 W
Catalog # 630-1255
Supplier:   MOTIC
Halogen bulbs
Catalog # 630-1255
Supplier:  MOTIC
Halogen bulbs
Halogen bulbs

Quartz halogen lamp, 6 V, 30 W

Price
29,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Quartz halogen lamp
  • Seq:
    601
  • Power:
    30 W
  • Voltage:
    6 V