ContactUsLinkComponent
FRIDGE TOTAL GROSS CAPACITY 92L TOTAL NET CAPACITY 86L CLASS A ENERGY RATING HEIGHT 630 X WIDTH 550 X DEPTH 620 MM INTEGRATED PREMIU M TABLE HEIGHT 1 * 1 items
  471-0363
 :  Liebherr
FRIDGE TOTAL GROSS CAPACITY 92L TOTAL NET CAPACITY 86L CLASS A ENERGY RATING HEIGHT 630 X WIDTH 550 X DEPTH 620 MM INTEGRATED PREMIU M TABLE HEIGHT 1 * 1 items
  471-0363
 :  Liebherr
Frequently Bought Together