Test kits, chlorine (DPD), CHECKIT®
Product Details & Documents
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
- Enables the use of large path lengths
- The mirror optics makes use of the view through the entire length of the cell
- Easy operation and exact reagent dosing
- Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
Delivery information: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.
Specifications
Specifications
- Pk:1 KIT
- Description:Chlorine test kit, DPD, (free, combined, total)
- Range:0 - 4 mg/l Cl<sub>2</sub>
Specifications
Accessories
Product Information
- Catalog #PkAvailabilityPrice
- CHECKIT® Comparator, path length 13,5 mm
Product Information
- Catalog #PkAvailabilityPrice
- CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path lengthCHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path lengthLOVI14550010Log in for availability1 * 10 ST - 41,30 €
Product Information
- Catalog #PkAvailabilityPrice
- CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path lengthCHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path lengthLOVI1455055Log in for availability1 * 5 ST - 24,40 €
Product Information
- Catalog #PkAvailabilityPrice
- CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path lengthCHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path lengthLOVI145510100Log in for availability1 * 100 ST - 342,00 €



