Orders LinkContactUsLinkComponent
Drip closures
Drip closures
Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%
Catalog # 215-3814
Supplier:   MUELLER KREMPEL
Drip closures
Catalog # 215-3814
Supplier:  MUELLER KREMPEL
Drip closures
Drip closures
Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%
Price
41,80 €
Log in for availability
1 * 100 ST
Product Details & Documents
Specifications
  • Description:
    Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%