Flasks, flat bottom with Quickfit® ground joint
150 ml, 105 mm, 34/34 NS
Catalog # 201-8437
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, clear, with short neck.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.
Specifications
- Socket size:34/34 NS
- Capacity:150 ml
- Height:105 mm
- Seq:10



