ContactUsLinkComponent
Aerometer, for plants, DIN12792, measuring range:1.000-1.210:0.002g/cm , without thermometer, length:325mm, reference temperature:20 °C, suitable for government verification. 1 * 1 items
Catalog # AMARH820258
Supplier:  Amarell
Aerometer, for plants, DIN12792, measuring range:1.000-1.210:0.002g/cm , without thermometer, length:325mm, reference temperature:20 °C, suitable for government verification. 1 * 1 items
Catalog # AMARH820258
Supplier:  Amarell
Frequently Bought Together