Orders LinkContactUsLinkComponent
Cell counters
Cell counters
Benchtop counter, 250×76×56 mm, 6
Catalog # HEATHS5360A
Supplier:   HEATHROW
Cell counters
Catalog # HEATHS5360A
Supplier:  HEATHROW
Cell counters
Cell counters
Benchtop counter, 250×76×56 mm, 6
Price
535,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These laboratory cell counters can measure cells such as basophils, eosinophils, myelocytes, juvenile, etc.
  • Each key records up to 999
  • Alarm at one hundred totals
  • Red and white buttons with grey housing
They can measure the number of cells such as Basophils (BASO), Eosinophils (EOSIN), Myelocytes (MYELO), Juvenile (JUVEN), Stab., Seg., Lymph., Mono., and total.
Specifications
  • Description:
    Benchtop counter, 250×76×56 mm
  • Array:
    6