Order Entry
Austria
Orders LinkContactUsLinkComponent
Mortars, without pestles

Mortars, without pestles

400 ml, 65 mm, 130 mm
Catalog # 410-0070
Mortars, without pestles
Catalog # 410-0070
Mortars, without pestles
Mortars, without pestles

400 ml, 65 mm, 130 mm

Price
79,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Porcelain with lip. Choice of glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

 : Pestle must be ordered separately.

Specifications
  • Ø int.:
    130 mm
  • Capacity:
    400 ml
  • Height:
    65 mm
  • Seq:
    07