Orders LinkContactUsLinkComponent
Volumetric Flasks, Class A
Volumetric Flasks, Class A
200 ml, 14/23, 0,15 ± ml
Catalog # WITG3.664.020.BL
Supplier:   witeg Labortechnik
Volumetric Flasks, Class A
Catalog # WITG3.664.020.BL
Supplier:  witeg Labortechnik
Volumetric Flasks, Class A
Volumetric Flasks, Class A
200 ml, 14/23, 0,15 ± ml
Price
32,40 €
Log in for availability
1 * 2 ST
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
  • DE-M marked according to the German calibration law
  • Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifications
  • NS:
    14/23
  • Capacity:
    200 ml
  • Seq:
    11
  • Tolerance:
    0,15 ± ml