Volumetric Flasks, Class A
5 ml, 10/19, 0,04 ± ml
Catalog # WITG3.664.015.BL
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifications
- NS:10/19
- Capacity:5 ml
- Seq:1
- Tolerance:0,04 ± ml