Orders LinkContactUsLinkComponent
Separating funnels, Squibb
100 ml, 19/26 NS
Catalog # WITG2.647.100
Supplier:   witeg Labortechnik
Separating funnels, Squibb
Catalog # WITG2.647.100
Supplier:  witeg Labortechnik
Separating funnels, Squibb
Separating funnels, Squibb
100 ml, 19/26 NS
Price
73,60 €
Log in for availability
1 * 2 ST
Product Details & Documents
Separating funnels, Squibb, made of borosilicate glass 3.3, with NS stopcock plugs and solid lass key. With PE-stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml
  • Seq:
    02