Order Entry
Austria
Orders LinkContactUsLinkComponent
Test tubes, disposable

Test tubes, disposable

20 ml, 150 mm, 16 mm
  SEMA1594
 :   SEMADENI
Test tubes, disposable
  SEMA1594
 :  SEMADENI
Test tubes, disposable
Test tubes, disposable

20 ml, 150 mm, 16 mm

 
29,70 €
   
1 * 100 ST
 

PS, clear.

  • Good thermal resistance
  • Good chemical resistance
  • Robust, economical

Universal tubes for a wide range of applications.

 
  • Ext.Ø:
    16 mm
  • Capacity:
    20 ml
  • Length:
    150 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.