Orders LinkContactUsLinkComponent
Certified colour reference standards
Certified colour reference standards
Pt-Co 100 mg Pt/l, certified under ISO 9001 quality system, Plastic bottle, 500 mL
Catalog # 633-0059
Supplier:   Lovibond Tintometer
Certified colour reference standards
Catalog # 633-0059
CAS Number:  
Certified colour reference standards
Certified colour reference standards
Pt-Co 100 mg Pt/l, certified under ISO 9001 quality system, Plastic bottle, 500 mL
CAS Number:  
Price
314,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
For routine calibration of colorimeters and verification of test data.
Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.
Documents
Specifications
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Pt-Co 100 mg Pt/l, certified under ISO 9001 quality system