Order Entry
Austria
Orders LinkContactUsLinkComponent
Double spatulas, micro | Avantor

Double spatulas, micro

130 mm, 5 mm
Artikelnr. 231-0467
Anbieter:   RSG
Double spatulas, micro
Artikelnr. 231-0467
Anbieter:  RSG
Double spatulas, micro
Double spatulas, micro

130 mm, 5 mm

Preis
36,70 €
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.

  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades

Dokumente

Spezifikationen
  • Length:
    130 mm
  • Blade width:
    5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.