Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Spatulas, micro

Round end, 150 mm, 5 mm
Catalog # RSGA087.035
Supplier:   RSG
Spatulas, micro
Catalog # RSGA087.035
Supplier:  RSG
Spatulas, micro
Spatulas, micro

Round end, 150 mm, 5 mm

Price
33,40 €
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel, 18/10. Micro spatulas with black plastic handle.

Specifications
  • Description:
    Round end
  • Length:
    150 mm
  • Blade width:
    5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.