Order Entry
Austria
Orders LinkContactUsLinkComponent
Spatulas, micro

Spatulas, micro

Bent spoon, 150 mm, 4 mm
Catalog # RSGA087.984
Supplier:   RSG
Spatulas, micro
Catalog # RSGA087.984
Supplier:  RSG
Spatulas, micro
Spatulas, micro

Bent spoon, 150 mm, 4 mm

Price
30,50 €
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel, 18/10. Micro spatulas with black plastic handle.

Specifications
  • Description:
    Bent spoon
  • Length:
    150 mm
  • Blade width:
    4 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.