Orders LinkContactUsLinkComponent
Forceps, straight
Forceps, straight
Fine tips, 145 mm
Catalog # 232-0075
Supplier:   RSG
Forceps, straight
Catalog # 232-0075
Supplier:  RSG
Forceps, straight
Forceps, straight
Fine tips, 145 mm
Price
62,30 €
Log in for availability
1 * 5 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
  • Serrated tips
  • With guide pin
Specifications
  • Type:
    Fine tips
  • Length:
    145 mm