Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, tower-tip, 110 mm
Catalog # 232-0188
Supplier:   RSG
Forceps
Catalog # 232-0188
Supplier:  RSG
Forceps
Forceps
Straight, tower-tip, 110 mm
Price
106,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
  • Very fine polished tips
Specifications
  • Type:
    Straight, tower-tip
  • Length:
    110 mm