Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
BNC connector for pH meters with BNC for example Hach, Orion, Mettler, 2 m
Catalog # SCOI285121221
Supplier:   SI Analytics
Electrode Cables
Catalog # SCOI285121221
Supplier:  SI Analytics
Electrode Cables
Electrode Cables
BNC connector for pH meters with BNC for example Hach, Orion, Mettler, 2 m
Price
76,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Electrode cables with a range of connectors to fit most pH meters.
Specifications
  • Description:
    BNC connector for pH meters with BNC for example Hach, Orion, Mettler
  • Length:
    2 m
  • Seq:
    17